VITLAB® Funnels, PP, BrandTech
Rugged, translucent polypropylene funnels are suitable for a variety of laboratory applications.
- Chemically resistant polypropylene
- 11 different sizes
Autoclavable at 121 °C (250 °F).
Ordering information: Larger sizes are available; contact your Avantor sales representative for information.
Rugged, translucent polypropylene funnels are suitable for a variety of laboratory applications.
- Chemically resistant polypropylene
- 11 different sizes
Autoclavable at 121 °C (250 °F).
Ordering information: Larger sizes are available; contact your Avantor sales representative for information.
- Catalog No:
- 83008-714
- 83008-716
- 83008-718
- 83008-720
- 83008-722
- 89140-952
- 89140-954
- 83008-725
- 89012-338
- 83008-726
- 83008-728
- Max. operating temperature:
- 125 °C (257 °F)
- 125 °C (257 °F)
- 125 °C (257 °F)
- 125 °C (257 °F)
- 125 °C (257 °F)
- 125 °C (257 °F)
- 125 °C (257 °F)
- 125 °C (257 °F)
- 125 °C (257 °F)
- 125 °C (257 °F)
- 125 °C (257 °F)
Product Family Options
- Overall LengthStem O.D.Top O.D.
- : 40894
Specifications
Cat. No.83008-714Max. operating temperature125 °C (257 °F) - : 41094
Specifications
Cat. No.83008-716Max. operating temperature125 °C (257 °F) - : 41194
Specifications
Cat. No.83008-718Max. operating temperature125 °C (257 °F) - : 41294
Specifications
Cat. No.83008-720Max. operating temperature125 °C (257 °F) - : 41394
Specifications
Cat. No.83008-722Max. operating temperature125 °C (257 °F) - : 41494
Specifications
Cat. No.89140-952Max. operating temperature125 °C (257 °F) - : 41594
Specifications
Cat. No.89140-954Max. operating temperature125 °C (257 °F) - 200 mm22 mm200 mmEach$265.73Pack of 6$251.55: 41794
Specifications
Cat. No.83008-725Max. operating temperature125 °C (257 °F) - : 41694
Specifications
Cat. No.89012-338Max. operating temperature125 °C (257 °F) - : 41894
Specifications
Cat. No.83008-726Max. operating temperature125 °C (257 °F) - : 41994
Specifications
Cat. No.83008-728Max. operating temperature125 °C (257 °F)