Nalgene® PassPort™ IP2 Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Excellent for small-scale inter-lab shipping of hazardous samples and chemicals, such as corrosives and cleaning compounds.
- Leakproof
- With polypropylene screw closures
The application of pressure or vacuum may result in product failure, damage to property, or personal injury.
Recommended for customers who are designing, assembling, and certifying their own combination packaging. Equipped with heavy-duty walls, bottles are designed to resist splitting and puncturing from shipping shocks; proven effective by industrial users. IP2 marking, material, volume, registration notch, and NALGENE name are molded into the bottle's bottom. Evaluated at 15 psi (103 kPa) per 49CFR 173.27 (c) (2) ICAO Technical Instructions Part 3-1; 1.1.6.1, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Ordering information: For replacement caps for 16057-169 and -170, see 16219-122 series.
Caution: Do not use bottle under pressure or vacuum.
Specifications
- Catalog No:
- 16057-164
- 16057-166
- 16057-160
- 16057-162
- 16057-170
- 16057-168
- 16057-169
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)