513 Basic Spectacle
Scratch-resistant/hardcoat, Clear, Blue
Catalog # 77393-756
Supplier: UNIVET OPTICAL TECHNOLOGIES
Supplier #: MDU5035S
Jump to:
Product Details & Documents
Colored temples in nonallergic rubber.
- Anti-scratch lens treatment
- Clear lens
The 513 Basic spectacle features a non-slip rubber nose pad and eyebrow protection.
Specifications
- Frame Color:Blue
- Lens Coating:Scratch-resistant/hardcoat
- Lens Tint:Clear