511 Basic Spectacles
511 Basic Spectacles
Certificates
511 Basic Spectacles
511 Basic Spectacles
2 Options Available Below
Product Details & Documents
The 511 Basic spectacle is designed with a frame made from non-allergic material.
  • Anti-fog and anti-scratch lens treatment
  • Adjustable temples
  • Integrated side shields
Temples are adjustable in length and inclination.
Product Family Options
Product Information
  • Lens Coating
    Lens Tint
    Frame Color
    Availability
    Price
  • Anti-fog, scratch-resistant
    Clear
    Black/orange
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $99.02
    $99.02
    Pack of 10
       $99.02
    Supplier Number: MDU5027SC
  • Anti-fog, scratch-resistant
    Clear
    Black
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $127.07
    $127.07
    Pack of 10
       $127.07
    Supplier Number: MDU5033S