Goggle Conversion Kit, 506U | Avantor
Goggle Conversion Kit, 506U
506U goggle conversion kit
Catalog # 77393-764
Fournisseur #:  MDU5040S
Goggle Conversion Kit, 506U
Catalog # 77393-764
Goggle Conversion Kit, 506U
Goggle Conversion Kit, 506U
506U goggle conversion kit
Price
$165.03
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Kit is designed to convert 506 Value spectacles into a hybrid spectacle (goggle).
  • Panoramic and wraparound lens for better side protection
  • Adjustable elastic band
Soft rubber gasket provides gentle support on the face.
Spécifications
  • Désignation:
    506U goggle conversion kit