546 Value Spectacles
546 Value Spectacles
546 Value Spectacles
Certificates
546 Value Spectacles
546 Value Spectacles
4 Options Available Below
Product Details & Documents
Extremely lightweight and modern spectacles with a sporty line.
  • Multiple lens treatments are available, including anti-fog, anti-scratch, and UV525
  • Wraparound polycarbonate lens
  • Integrated side shields
546 Value spectacles feature temples equipped with rubber terminals to guarantee excellent comfort, as well as a non-slip rubber nose pad. The UV525 protects eyes during curing light usage. Smoke lenses can help to reduce anxiety.
Product Family Options
Informations sur le produit
  • Lens Coating
    Lens Tint
    Frame Color
    Availability
    Price
  • Anti-fog, scratch-resistant
    Clear
    Black/orange
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $198.03
    $198.03
    Pack of 10
       $198.03
    Supplier Number: MDU5028SC
  • Anti-fog, scratch-resistant, UV525
    Orange
    Black/orange
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $198.03
    $198.03
    Pack of 10
       $198.03
    Supplier Number: MDU5029SC
  • Anti-fog, scratch-resistant
    Clear
    Black/green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $198.03
    $198.03
    Pack of 10
       $198.03
    Supplier Number: MDU5038SC
  • Anti-fog, scratch-resistant
    Smoke
    Black/green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $198.03
    $198.03
    Pack of 10
       $198.03
    Supplier Number: MDU5030S