6×1 X-Gen Goggle
Clear, Gunmetal grey/green
Catalog # 77393-770
Supplier: UNIVET OPTICAL TECHNOLOGIES
: MDU6005S
Jump to:
Product Details & Documents
6×1 X-Gen goggles feature a broad lens for a wide field of view with zero distortions.
- Lens treatment: anti-fog and anti-scratch (double coating), plus UV380
- Gunmetal/green frame color
- Clear lens
Goggles are designed with an internal soft rubber to ensure a comfortable fit with no inconvenient high pressure points. They feature an interchangeable system between temples and elastic band (included in the package). The temples have rubber terminals and are adjustable in length and inclination. The lens is treated with VPlus anti-fog and anti-scratch enhancement.
: Certified according to EN 166 standard, level 3 and 4 (EU) for protection against droplets, splashes, and large dust particles.
Spécifications
- Frame Color:Gunmetal grey/green
- Lens Tint:Clear