VWR® Adjustable Spray Bottle | Avantor
VWR® Adjustable Spray Bottle
237 ml (8 oz.)
Catalog # 23609-182
Supplier:   VWR International
VWR® Adjustable Spray Bottle
Catalog # 23609-182
Supplier:  VWR International
VWR® Adjustable Spray Bottle
VWR® Adjustable Spray Bottle
237 ml (8 oz.)
Price
$86.60
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 3
Product Details & Documents
The bottle's adjustable sprayer permits a range of positioning from fine mist to large stream delivery to accommodate a variety of washing applications.
  • HDPE and polypropylene construction
  • Chemical resistant and unbreakable
  • Instant-off release eliminates messy dripping
The chemical-resistant and unbreakable bottle is constructed of HDPE and the sprayer is made from polypropylene. The pump trigger is comfortable for one-handed operation and features an instant-off release that eliminates messy dripping. Closed position on sprayer maintains solution purity. The entire wash bottle may be gas or chemically sterilized. Ideal for hot, cold, aqueous, and solvent-based solutions.
Spécifications
  • Capacité:
    237 ml (8 oz.)