ContactUsLinkComponent
Cloth Parts Bags with Single Drawstring, Associated Bag
Catalogue # 47745-018
Fournisseur:  Associated Bag
undefined
Cloth Parts Bags with Single Drawstring, Associated Bag
Catalogue # 47745-018
Fournisseur:  Associated Bag
Supplier Number:  29-14
Spécifications
  • Size:
    6×10"
  • Cat. no.:
    47745-018
  • Supplier No.:
    29-14
Spécifications
À propos de cet article
100% cotton, including drawstring ties.
  • Breathe and absorb moisture to prevent corrosion
Store and ship plumbing supplies, electrical switches, hardware, and more. Mailing bags have preprinted address tag attached to the bottom.