Sterileze® Gas Sterilization Pouches, Amcor Flexibles
Fournisseur: Amcor Flexibles Healthcare
Certificats
À propos de cet article
These clear plastic pouches feature a breathable Tyvek® vent that prevents blowup during sterilization cycles and allows fast aeration.
The Tower-Tear® snap-open feature provides quick, easy access to pouch contents. The pouches may be heat-sealed or taped.
Moisture and puncture-resistant pouches offer an excellent bacterial barrier. Pouches are 3 mil thick.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together