VWR® Red Wash Bottles, LDPE
Fournisseur: VWR International
Certificats
À propos de cet article
Low-density polyethylene bottles molded in bright red provide easy identification of hazardous materials.
- Vibrant red color for quick identification
- Ideal for dangerous solvents and solutions
Durable, flexible, chemical-resistant, and easy to clean. Excellent pliability allows comfortable squeeze flow control. Has one-piece molded polypropylene cap and nozzle. Dispense a single drop or a needle sharp stream. If a coarser flow is desired, dispensing tip can be trimmed.
Frequently Bought Together