Precision Clean® II Class 100 Cleanroom Bags, PPC Flexible Packaging
Supplier: Fisher Container
Certificates
About this item
These bags are ideal for packaging applications requiring ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom conditions.
They are made of clear polyethylene. Choose from 2, 4, or 6 mil thickness.
Frequently Bought Together