Nalgene® Large Size Funnels, Polyethylene, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Ideal for transferring liquids
Funnels have both internal and external ribbing for air release. High-density polyethylene.
Frequently Bought Together