pH Test Strips with Pads
Supplier: Precision Laboratories
Jump to:
À propos de cet article
The strips have three pads for enhanced accuracy. Results are available in seconds.
- pH 1 to 14 test strips are wide-range strips measuring in steps of 1.0 pH units
- pH 0 to 7 test strips measure the entire range of acid from a pH 0.0 to a neutral pH 7.0 in steps of 0.5 pH units
- pH 7 to 14 test strips are used to test neutral to very alkaline solutions in increments of 0.5 pH units
- pH 2 to 9 test strips measure a wide range of both acidic and alkaline solutions in steps of 0.5 pH units
- pH 4.5 to 10 test strips are medium range strips ideal for testing slightly acidic to slightly alkaline solutions in steps of 0.5 pH units
The strips have three pads for accuracy with results available in seconds. The strips are available in five ranges 0 to 14, 0 to 7, 7 to 14, 2 to 9 and 4.5 to 10.
10 Options Available Below
Product Details & Documents
The strips have three pads for enhanced accuracy. Results are available in seconds.
- pH 1 to 14 test strips are wide-range strips measuring in steps of 1.0 pH units
- pH 0 to 7 test strips measure the entire range of acid from a pH 0.0 to a neutral pH 7.0 in steps of 0.5 pH units
- pH 7 to 14 test strips are used to test neutral to very alkaline solutions in increments of 0.5 pH units
- pH 2 to 9 test strips measure a wide range of both acidic and alkaline solutions in steps of 0.5 pH units
- pH 4.5 to 10 test strips are medium range strips ideal for testing slightly acidic to slightly alkaline solutions in steps of 0.5 pH units
The strips have three pads for accuracy with results available in seconds. The strips are available in five ranges 0 to 14, 0 to 7, 7 to 14, 2 to 9 and 4.5 to 10.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: PH0007-3-1B-50
- Supplier Number: PH0007-3-1V-100
- Supplier Number: PH0114-3-1B-50
- Supplier Number: PH0114-3-1V-100
- Supplier Number: PH2090-3-1B-50
- Supplier Number: PH2090-3-1V-100
- Supplier Number: PH4510-3-1B-50
- Supplier Number: PH4510-3-1V-100
- Supplier Number: PH0714-3-1B-50
- Supplier Number: PH0714-3-1V-100