BIOXP® Select Plasmid AMP
Supplier:   TELESIS BIO
BIOXP® Select Plasmid AMP
Supplier:  TELESIS BIO
Certificates
BIOXP® Select Plasmid AMP
BIOXP® Select Plasmid AMP
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
BioXp Select plasmid amplification kits contain reagents for isothermal amplification of plasmid DNA.
  • Input: Plasmids ranging from 1.8 to 37 kb
  • Yield: >10 µg/well
  • Output format: Branched concatemers
  • Compatible with BioXp 3250
Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.
Product Family Options
Product Information
  • Désignation
    No. of Reactions
    Availability
    Price
  • Kit for BioXp 3250
    8
    Log in for availability
    Log in for availability
    Each
    Each - $1,981.98
    $1,981.98
    Each
       $1,981.98
    Supplier Number: BX-SEL-PLSAMP-08
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Kit for BioXp 3250
    16
    Log in for availability
    Log in for availability
    Each
    Each - $3,963.97
    $3,963.97
    Each
       $3,963.97
    Supplier Number: BX-SEL-PLSAMP-16
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Kit for BioXp 3250
    24
    Log in for availability
    Log in for availability
    Each
    Each - $5,945.95
    $5,945.95
    Each
       $5,945.95
    Supplier Number: BX-SEL-PLSAMP-24
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.