SP Bel-Art Clavies™ Bag Clips, Bel-Art Products, a part of SP
12000-072
- Cat. no.:12000-072
- W×D×H:11 mm (⁷/₁₆")
- Supplier No.:131900100
Stiff plastic design provides secure closure for autoclave bags yet allows release of internal pressure that develops during autoclave cycle.
- Disposable clip is easily closed and opened with one hand
- Secure closure
Suitable for benchtop or small autoclave bags up to 48.3×58.4 cm (19×23"); use with larger bags is not recommended.