Tapered Five-Neck Flasks, Jacketed, Ace Glass
À propos de cet article
Flasks have tapered walls to allow continuous operation with smaller volumes
The full jacket has two barbed side tubes for hoses. These five-neck flasks have three [ST] outer joints and two #7 Ace-Thred necks. The outer joints are tooled with reinforcement rings for added strength and stability. Two of the side necks are in-line with the center neck. The remaining side necks are placed in front of the in-line necks and are internally threaded to accommodate thermometers or bleed tubes up to 7mmO.D. Supplied with two #7 nylon bushings and FETFE O-rings.
6 Options Available Below
Product Details & Documents
Flasks have tapered walls to allow continuous operation with smaller volumes
The full jacket has two barbed side tubes for hoses. These five-neck flasks have three [ST] outer joints and two #7 Ace-Thred necks. The outer joints are tooled with reinforcement rings for added strength and stability. Two of the side necks are in-line with the center neck. The remaining side necks are placed in front of the in-line necks and are internally threaded to accommodate thermometers or bleed tubes up to 7mmO.D. Supplied with two #7 nylon bushings and FETFE O-rings.
Product Family Options
Informations sur le produit
- CapacitéCenter Neck [ST] JointSide Neck ST JointsSide Tube I.D.AvailabilityPrice
- 125 ml14/2014/209.5 mm (3/8")Log in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,747.97$1,747.97Prix promotionnel pour les nouveaux clientsEach$1,747.97Supplier Number: 6960-09
- 250 ml24/4024/4011.1 mm (7/16")Log in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,978.28$1,978.28Prix promotionnel pour les nouveaux clientsEach$1,978.28Supplier Number: 6960-18
- 250 ml29/4224/4011.1 mm (7/16")Log in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,978.28$1,978.28Prix promotionnel pour les nouveaux clientsEach$1,978.28Supplier Number: 6960-20
- 500 ml24/4024/4012.7 mm (1/2")Log in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,378.99$2,378.99Prix promotionnel pour les nouveaux clientsEach$2,378.99Supplier Number: 6960-35
- 500 ml29/4224/4012.7 mm (1/2")Log in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,378.99$2,378.99Prix promotionnel pour les nouveaux clientsEach$2,378.99Supplier Number: 6960-37
- 50 ml14/2014/209.5 mm (3/8")Log in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,747.97$1,747.97Prix promotionnel pour les nouveaux clientsEach$1,747.97Supplier Number: 6960-02