KIMAX® Volumetric Flask with [ST] Glass Stopper, Class A, to Contain and Deliver, Kimble Chase
Catalog # 89002-062
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:100 ml
- Cat. no.:89002-062
- ST Stopper No.:13
- Tolerance:±0.08 ml to contain, ±0.15 ml to deliver
- Supplier No.:28026-100
Specifications
About this item
Volumetric flask is calibrated both to contain and to deliver
Flask features a marking spot, [ST] ground glass stopper, and graduation ring blasted on the neck. Designed from ASTM Specification E288, Class A unserialized requirements.