PYREX® VISTA™ Volumetric Flasks with Snap Caps, Class B, Corning
: Corning
Made of PYREX® glass, with PE snap-cap.
- Heavy duty necks
- Graduation line is sharp and permanent
- Inscriptions easy to read
Designed for use by customers that do not require the precision accuracy of our Class A capacity flasks. The shape, body and wall are the same as regular flasks but display the large blue marking spot and designation 'PYREX VISTA' unique to the PYREX VISTA line. Capacity tolerances are twice those specified for Class A volumetric ware.
Heavy-duty necks are tooled for HDPE snap-caps, which are included with all sizes. Flat bottom design with uniform wall thickness ensures reliable, long-lasting use. Reusable.
- Catalog No:
- 89090-306
- 89090-300
- 89090-344
- 89090-382
- 89090-356
- 89090-118
- 89090-332
- 89090-394
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Frequently Bought Together