VWR® Talking Timer
0.01%, 6.4×8.3×1.3 cm (2½×3¼×½")
  62379-565
VWR® Talking Timer
  62379-565
VWR® Talking Timer
VWR® Talking Timer
0.01%, 6.4×8.3×1.3 cm (2½×3¼×½")
$97.76
Each
Innovative timer talks, times up, counts down, and tells time-of-day with traceable precision.
  • Traceable®
  • Timer talks, times up, and counts down
  • Magnetic backing for convenient mounting
Unique 'thru-zero' feature allows alarming, counting up, and counting down from 23 hours, 59 minutes, 59 seconds, with one-second resolution. Large, 1.3 cm (¹/₂") high display and LCD digits continuously show time remaining. During countdown, voice reporting frequency is every hour until the last hour is reached, then every 10 minutes until 1 minute is reached, then every 10 seconds until 10 seconds is reached, then every second. Timer also announces elapsed time since alarming. A time-out mode stops the timer for 'off' periods for both up and down timing. In clock mode, timer announces time-of-day. Magnetic backing allows placement of timer on lab refrigerators or incubators. Stand timer on a lab bench using flip-open easel, or clip to a clipboard using the spring fastener.
 : A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
 : Unit is supplied with two 1.5 V silver-oxide batteries for continuous two-year use.
  • W×H×D:
    6.4×8.3×1.3 cm (2½×3¼×½")
  • Accuracy:
    0.01%