Volumetric Flasks with [ST] Glass Stopper, Class A, Amber, Hirschmann
Flasks feature amber-colored glass for the protection of light-sensitive contents, a fine white graduation line, and marking spot
Meet ASTM E288, Class A requirements.
Graduated to contain. Flasks include a [ST] glass stopper.
Frequently Bought Together