Staticide® Antistatic Floor Stripper, ACL
Staticide® Antistatic Floor Stripper, ACL
Supplier:   ACL Staticide
Staticide® Antistatic Floor Stripper, ACL
Supplier:  ACL Staticide
Certificates
Staticide® Antistatic Floor Stripper, ACL
Staticide® Antistatic Floor Stripper, ACL
3 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
This high-performance floor stripper removes multiple coats of wax or synthetic floor finishes
The dual action quickly and effectively lifts coatings and removes embedded dirt. The result is a completely clean and finish-ready surface. Non-streaking, low-foaming stripper will not damage or dull flooring when used according to flooring manufacturer’s recommendations. Effective in hard or soft water. Non-ammoniated and biodegradable.
Product Family Options
Product Information
  • Taille
    Packaging
    Availability
    Price
  • 3.8 L (1 gal.)
    Bottle
    Log in for availability
    Log in for availability
    Case of 4 (4l)
    Case of 4 (4l) - $323.77
    $323.77
    Case of 4 (4l)
       $323.77
    Supplier Number: 4010-1
  • 18.9 L (5 gal.)
    Pail
    Log in for availability
    Log in for availability
    Each
    Each - $291.17
    $291.17
    Each
       $291.17
    Supplier Number: 4010-5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 204 L (54 gal.)
    Drum
    Log in for availability
    Log in for availability
    Each (54US Gal)
    Each (54US Gal) - $2,688.74
    $2,688.74
    Each (54US Gal)
       $2,688.74
    Supplier Number: 4010-2