Static Free™ Mat and Benchtop Reconditioner, ITW Chemtronics
Catalog # 15145-435
Supplier: ITW Chemtronics
Specifications
- Size:473 mL (16 oz.)
- Cat. no.:15145-435
- Packaging:Trigger Spray Bottle
Specifications
About this item
Static-dissipative cleaner and reconditioner for ESD mats, benchtops, and production assembly surfaces can rejuvenate dead spots on ESD mats
This nonflammable, nonabrasive cleaner is noncorrosive and chloride- and amine-free. Safe to use on any surface not affected by water; will not dry out ESD mats.