Staticide® Antistatic Solution, ACL
Staticide® Antistatic Solution, ACL
Staticide® Antistatic Solution, ACL
Supplier:  ACL Staticide
Certificates
Staticide® Antistatic Solution, ACL
Staticide® Antistatic Solution, ACL
2 Options Available Below
Product Details & Documents
Easy-to-use topical antistatic solution for long-term static control
Choose from pour-top gallons or ready-to-use quart bottles with sprayer. Cleanroom formula is individually bagged to prevent contamination.
Apply to floors, walls, ceilings, fixtures, work surfaces, carpets, and more. Effective in PC-board solder fault reduction programs; used in high sensitivity electronic and EDP operations. Nontoxic and nonflammable. Completely biodegradable
Certifications: Cleanroom formula is ISO Class 5 (FED-STD-209E Class 100/M3.5) approved. Meets MIL-B-81705B and NFPA-56A electrostatic decay criteria.
Documents
Product Family Options
Product Information
  • Désignation
    Taille
    Availability
    Price
  • General-Purpose
    946 mL (1 qt.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $74.58
    $74.58
    Log in for availability
    Case of 12 (1/ Itm/Bx)
    Case of 12 (1/ Itm/Bx) - $894.96
    $894.96
    Each(1/Itm/Bx)
       $74.58
    Case of 12 (1/ Itm/Bx)
       $894.96
    Supplier Number: 2003
  • General-Purpose
    3.8 L (1 gal.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $168.77
    $168.77
    Log in for availability
    Case of 4 (1/ Itm/Bx)
    Case of 4 (1/ Itm/Bx) - $675.08
    $675.08
    Each(1/Itm/Bx)
       $168.77
    Case of 4 (1/ Itm/Bx)
       $675.08
    Supplier Number: 2001