Mat and Tabletop Cleaner, ACL
À propos de cet article
Formulated to clean tabletops, antistatic mats, and workstations
Removes flux residues, oil, grease, and inorganic contaminants. Perfect for assembly areas where sensitive electronic components are handled. Maintains original surface resistivity of antistatic mats. Nonflammable and safe for any surface.
2 Options Available Below
Product Details & Documents
Formulated to clean tabletops, antistatic mats, and workstations
Removes flux residues, oil, grease, and inorganic contaminants. Perfect for assembly areas where sensitive electronic components are handled. Maintains original surface resistivity of antistatic mats. Nonflammable and safe for any surface.
Documents
Product Family Options
Informations sur le produit
- PackagingTailleAvailabilityPrice
- Spray Bottle473 mL (16 oz.)Each(1/Itm/Bx)$56.02Case of 12 (1/ Itm/Bx)$434.07Supplier Number: 6001
- Container3.8 L (1 gal.)Each(1 US Gal)$139.61Case of 4 (1 US Gal)$414.36Supplier Number: 6002