VWR® Traceable® Thermometer/Clock/Humidity Monitor
62344-734
- Description:Thermometer, 12/24 Hour Clock, and Humidity Monitor
- Environmentally Preferable:
- Temperature accuracy:±1°C
- Temperature resolution:0.1°
- Humidity resolution:1% RH
- Humidity accuracy:±5% RH mid-range, otherwise ±8%
- W×D×H:5.7×1.3×10.8 cm (21/4×1/2 ×41/4")
- Cat. no.:62344-734
- Range:0...50 °C
20...90% RH - Weight:71 g
Unit simultaneously displays ambient temperature, humidity, and time of day.
- Traceable®
- High/low temperature and humidity memory
- Easy-to-read display
Ideal for use in incubators, storerooms, drying chambers, hoods, cleanrooms, and environmental cabinets.
Memory recalls highest and lowest temperature and humidity readings. Clock displays a.m./p.m. or 24-hour military time to the exact minute with 0.01% accuracy.
: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
: Supplied with one-year AAA battery, built-in stand, and wall mount.