Lisle Inspector’s Gloves, Protective Industrial Products
Supplier: PIP
Certificates
About this item
Premium, light weight cotton lisle inspection ambidextrous gloves with unhemmed cuff.
- Constructed in a two piece, reversible pattern using 100% cotton fabric
- Designed to minimize contamination of products from human hands
- Can be used as liners under disposable and unsupported liquid proof gloves
Specifications
- Catalog No:
- 32932-212
- 32932-261
- 32932-210
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous