KIMAX® Volumetric Flasks, Cassia, Class A, Kimble Chase
Catalogue # 29630-000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Capacity:110 ml
- Cat. no.:29630-000
- ST Stopper No.:13
- Supplier No.:28066-110
Spécifications
À propos de cet article
Calibrated to contain
For assaying essential oils according to methods of the U.S. Pharmacopoeia and Essential Oil Association of the U.S.A. White ceramic enamel scale. Supplied with a [ST] glass stopper and marking spot.