KIMAX® Volumetric Flasks with Snap Cap, Class A, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Calibrated to contain
Designed from ASTM Specification E288 and with Class A accuracy but without serial number.
Supplied with a polyethylene snap cap.
Frequently Bought Together