Score-Break Ampules, KIMBLE®, DWK Life Sciences
Score-break ampules made from borosilicate glass.
- Short stem ampules are tip-sealed
- Trimmed stems are pull-sealed
Designed with a score-mark on the constriction breakpoint for opening ease
Frequently Bought Together