Wallcur® Practi-Sufentanil
Product Details & Documents
Made for training.
- Contains no medication
- Contents are safe for use with most simulators
Practi-Sufentanil™ 100 mcg/2 ml Amp simulates the narcotic sufentanil (Sufenta®), commonly used to treat pain or used along with anesthesia during surgery or childbirth. Each 2 ml distilled water ampule is labeled Practi-Sufentanil™ 100 mcg/2 ml, designed to aid in a complex learning experience requiring critical thinking when calculating, preparing, and administering dosages.
Manufactured for safety and consistency, each Practi-Ampule is chemically scored at the neck for ease in breakage without injury. Health occupational curricula, such as nursing, pharmacy tech, emergency tech, and surgical tech programs will find the Practi-Sufentanil Ampule the most authentic practice available. Snap, aspirate, calculate, measure, and administer fractional dosages from an ampule safely and authentically.
Delivery information: These products require a release form for purchase. If your account does not have one on file, the item cannot ship until release documentation is received.
Caution: For training purposes only. Not for human or animal use.
Spécifications
- Désignation:Practi-Sufentanil (100 mcg/2 ml)
- Includes:Distilled water