Flat-Bottom Three-Neck Flasks, Vertical Necks, Ace Glass
Fournisseur: Ace Glass
Certificats
À propos de cet article
Flat-bottom flask can sit directly on heater or stirrer without other support
The [ST] 24/40 outer joints are tooled with reinforcement rings for added strength and stability.
Ordering information: Vertical neck flasks can be ordered for angled neck capacities; contact your Avantor sales representative.
Frequently Bought Together