NEXTFLEX™ Universal Blockers
NEXTFLEX™ Universal Blockers
NEXTFLEX™ Universal Blockers
NEXTFLEX™ Universal Blockers
The NEXTFLEX™ universal blockers block non-specific hybridization, enhancing specificity and reducing sequencing costs. They are compatible with all ligation and tagmentation workflows for Illumina®
  • Effective in blocking single-index or dual-index libraries
  • Compatible with all NEXTFLEX™ barcodes and all other ligation-based and tagmentation-based workflows for Illumina® sequencing platforms
  • Simplifies and reduces the cost of target capture without sacrificing performance
  • 96 barcode blockers available
During target enrichment, adapters can interact with the complementary adapter sequence strand of another library molecule. The NEXTFLEX™ universal blockers are designed to prevent this concatemerization of library molecules. By blocking this complementary adapter sequence interaction, the number of on-target reads improves dramatically and the depth of enrichment increases. The NEXTFLEX™ universal blockers are designed for use with indexed adapters for Illumina sequencing platforms. They are effective in blocking single-indexed or dual-indexed libraries, without being affected by index length or the presence of a unique molecular index (UMI). The NEXTFLEX Universal Blockers are compatible with ligation-based workflows, such as NEXTFLEX™ or Illumina trusequence library prep kits. They are also compatible with tagmentation-based workflows, such as Illumina Nextera library prep kits.
 : Manufactured in ISO 9001 facility.
 : For research use only. Not for use in diagnostic procedures.
Product Family Options
  • Désignation
    No. of Reactions
  • NEXTFLEX™ universal blockers
    8
    Each
    Each - $187.29
    $187.29
    Each
       $187.29
     : NOVA-5143231
  • NEXTFLEX™ universal blockers
    8
    Each
    Each - $151.06
    $151.06
    Each
       $151.06
     : NOVA-5143231
  • NEXTFLEX™ universal blockers
    48
    Each
    Each - $316.36
    $316.36
    Each
       $316.36
     : NOVA-5143232
  • NEXTFLEX™ universal blockers
    96
    Each
    Each - $497.35
    $497.35
    Each
       $497.35
     : NOVA-5143233