NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits | Avantor
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
Certificates
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
3 Options Available Below
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The NEXTFLEX® Rapid XP V2 DNA-sequence kit offers a simplified DNA-seq library prep workflow for Illumina® platforms. Libraries produced using this kit are normalized during library construction.
  • Fully automated normalization and fragmentation
  • Enzymatic fragmentation with end-repair and A-tailing in one reaction
  • Highly efficient ligation with low adapter dimer formation
  • Efficient: Automated on the Sciclone®  and Zephyr®  NGS workstations for mid-high throughput automation and now on the BioQule™ NGS System for low throughput labs
The NEXTFLEX® Rapid XP V2 DNA-seq kit includes proprietary NEXTFLEX® normalization beads, which provide a consistent mass and sequencing cluster density for all samples in a library pool. This shortens the time needed for quantification and pooling preparation for sequencing in a high throughput lab by up to 3 hours per 96 samples. The NEXTFLEX  Rapid XP V2 DNA-seq kit offers a streamlined workflow with highly efficient ligation, generating low amount of adapter dimers even at low input concentrations.
The NEXTFLEX Rapid XP V2 DNA-Seq kit is compatible with multiple liquid handlers, including the Sciclone G3 NGSx workstation, Sciclone G3 NGSx iQ™ workstation, and Zephyr NGS workstation. For low throughput automation, the NEXTFLEX  Rapid XP V2 DNA-Seq kit is also now automated on the BioQule™ NGS System which simplifies both library prep and quantitation. This low-cost, benchtop instrument delivers libraries ready to load into your sequencer with only 15 minutes of hands-on time. 
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Reactions
    Availability
    Price
  • NEXTFLEX™ Rapid XP V2 DNA-sequence kit
    8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $302.53
    $302.53
    Each
       $302.53
    La quantité minimale de commande est de 1
    Supplier Number: NOVA-5149-21
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • NEXTFLEX™ Rapid XP V2 DNA-sequence kit
    48
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,797.04
    $1,797.04
    Each
       $1,797.04
    La quantité minimale de commande est de 1
    Supplier Number: NOVA-5149-22
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • NEXTFLEX™ Rapid XP V2 DNA-sequence kit
    96
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,564.34
    $3,564.34
    Each
       $3,564.34
    La quantité minimale de commande est de 1
    Supplier Number: NOVA-5149-23
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.