PG-Seq™ Rapid Kit V2 and Index Primers
The PG-Seq™ Rapid kit V2 has been developed to analyze picogram quantities of DNA (single/multi-cells or low template DNA) from an embryo biopsy for preimplantation genetic testing.
- Updated, streamlined, simple PGT protocol
- Accurate copy number detection
- Flexible kit format
The PG-Seq™ Rapid kit V2 has been developed to analyze picogram quantities of DNA (single/multi-cells or low template DNA) from an embryo biopsy for preimplantation genetic testing. The PG-Seq™ Rapid kit v2 utilizes whole genome amplification (WGA) and next generation sequencing (NGS) technology to accurately screen all 24 chromosomes for whole chromosome aneuploidy and sub-chromosomal abnormalities.
: Manufactured in ISO 9001 facility.
: PG-seq™ Index primers must be ordered with PG-seq kits.
: For research use only. Not for use in diagnostic procedures.
Frequently Bought Together