Derma Free® Vinyl Gloves, Microflex®
These ambidextrous and nonsterile gloves are ideal for food service, food processing, or any application where personal hygiene is critical.
- Powder-free
- Beaded cuffs
- Ambidextrous
- Nonsterile
- Synthetic vinyl construction
- Thickness: 3.9 mil (finger)
Gloves are thicker for added protection and have a smooth surface for more natural tactile sensitivity.
Medical-exam-grade gloves are made of 100% synthetic vinyl.
: Caution Information: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
- Catalog No:
- CA13713-008
- CA13713-010
- CA13713-006
- CA13713-004
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- Finger thickness:
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- Palm thickness:
- 4.3 mil
- 4.3 mil
- 4.3 mil
- 4.3 mil
Frequently Bought Together