LDPE Cubitainer® with LDPE Foam-lined Closure, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Cubitainers™ are designed for the efficient storage and transport. Recommended for collecting water samples and other low viscosity liquids to be analysed for inorganic trace analytes.
- Light weight and space-efficient solution for remote locations where motorised vehicle transportation options are limited
- Collapsible when empty for efficient storage and transport, or for efficient packing into small spaces like back packs for remote sampling expeditions
- Unprocessed container comes assembled and ready for own cleaning procedure
- With white LDPE-lined PP cap (assembled)
Certified containers meet or exceed EPA performance-based standards for metals, cyanide and fluoride. It includes in the case and barcoded with lot number for traceability, unattached analysis labels.
Ordering information: Supplied with polyfoam-lined polypropylene caps.
Frequently Bought Together