Polycarbin® LTS® Compatible Refill Towers
À propos de cet article
Polycarbin's LTS® compatible refill towers deliver the precision and accuracy you expect, with a fraction of the carbon footprint.
- Wafer and rack made from 100% circular economy, post-consumer recycled content
- Smaller carbon footprint compared to other refill and rack systems
- Easy to transfer refills into polycarbin racks
- Certified DNase, RNase and pyrogen free
- Non-sterile (autoclavable)
Polycarbin's LTS® compatible refill system is designed to deliver an exceptional product that is less carbon and plastic-intensive. Refill tower housing is manufactured using post-consumer recycled content, and are shrink wrapped and packaged in an easy to recycle cardboard sleeve. Pipette tips do not contain recycled content and are made from virgin PP resin.
Certifications: Certified sterile and DNase, RNase and pyrogen free. Manufactured in an ISO 9001 compliant facility.
Ordering information: Note that Polycarbin's LTS® compatible refill towers do not come with racks (bases). These can be ordered separately by the sleeve. Please see part numbers 76635-484, 76635-488 and 76635-492.
3 Options Available Below
Product Details & Documents
Polycarbin's LTS® compatible refill towers deliver the precision and accuracy you expect, with a fraction of the carbon footprint.
- Wafer and rack made from 100% circular economy, post-consumer recycled content
- Smaller carbon footprint compared to other refill and rack systems
- Easy to transfer refills into polycarbin racks
- Certified DNase, RNase and pyrogen free
- Non-sterile (autoclavable)
Polycarbin's LTS® compatible refill system is designed to deliver an exceptional product that is less carbon and plastic-intensive. Refill tower housing is manufactured using post-consumer recycled content, and are shrink wrapped and packaged in an easy to recycle cardboard sleeve. Pipette tips do not contain recycled content and are made from virgin PP resin.
Certifications: Certified sterile and DNase, RNase and pyrogen free. Manufactured in an ISO 9001 compliant facility.
Ordering information: Note that Polycarbin's LTS® compatible refill towers do not come with racks (bases). These can be ordered separately by the sleeve. Please see part numbers 76635-484, 76635-488 and 76635-492.
Product Family Options
LTS® compatible refill towers
Informations sur le produit
- VolumeSterileLongueurTip ColorPackedAvailabilityPrice
- Supplier Number: PC01-06-06R
- Supplier Number: PC01-05-06R
- Supplier Number: PC01-04-06R