NEBNext® Primers for Epigenetics (Unique Dual Index Set 3)
Catalog # CA77441-302
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:NEBNext® primers for epigenetics (Unique Dual Index Set 3)
- Size:96 reactions
- Cat. no.:CA77441-302
Specifications
About this item
The NEBNext Primers for Epigenetics contain unique dual index primers optimized for E5hmC-seq libraries.
- Optimized for NEBNext Epigenetics workflows
This 96 reaction kit contains premixed i5 and i7 8-base index primers, and each well of a 96-well plate contains an i5 and i7 primer pair.