NEBNext® Enzymatic 5hmC-seq Conversion Modules
Supplier: New England Biolabs (NEB)
Certificates
About this item
The NEBNext Enzymatic 5hmC-seq Conversion Module contains the reagents required to enzymatically modify and enable specific detection of 5-hydroxymethylcytosine (5hmC).
- Specific 5hmC conversion
- Enzyme-based conversion minimizes DNA damage
- High-efficiency conversion
- 0.1 to 200 ng input range
The user-friendly workflow enables 5hmC identification from 0.1 to 200 ng of input DNA.
Frequently Bought Together