NEBNext® UltraExpress™ FS DNA Library Prep Kits
Supplier: New England Biolabs (NEB)
Certificates
About this item
The NEBNext UltraExpress FS DNA Library Prep kit is the latest generation of NEBNext DNA library prep, with a fast, streamlined workflow, including a fragmentation step.
- Fast workflow (<2 hours) with integrated enzymatic fragmentation step
- Fewer steps and consumables
- Fewer cleanups
- Single protocol for all input amounts
- High-quality libraries from a wide input range (10 to 200 ng intact DNA)
In under 2 hours, the protocol enables the creation of high yield, high quality libraries, from a broad input amount range of intact DNA, while generating less plastic waste. Further, with simplicity at the forefront, the kit features a single protocol for all input amounts, making NEBNext UltraExpress DNA Library Prep well suited for all throughputs.
Frequently Bought Together