Battery Hydrometer and Syphon Set
Battery Hydrometer and Syphon Set
Supplier:   Heathrow Scientific
Battery Hydrometer and Syphon Set
Certificates
Battery Hydrometer and Syphon Set
Battery Hydrometer and Syphon Set
3 Options Available Below
Product Details & Documents
Our battery acid hydrometer set is used in many industrial settings including fleet maintenance, automotive services, solar-powered system maintenance, and general industrial applications.
  • The rubber parts are made of nitrile rubber, which is chemically inert and durable
  • Calibrated for use at 77 °F
A reinforced box provides storage and is compartmented to eliminate breakage.
Delivery information: The set includes a specific gravity hydrometer and glass syphon tube with a rubber bulb, nozzle, and clamp.
Product Family Options
Informations sur le produit
  • Désignation
    Range
    Longueur
    Subdivisions
    Availability
    Price
  • Hydrometer only
    1.150 - 1.300 (sp. gravity)
    220 mm (8.7")
    0.005
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $93.26
    $93.26
    Prix promotionnel pour les nouveaux clients
    Each
       $93.26
    Supplier Number: 6605-5H
  • Hydrometer and syphon set
    1.150 - 1.300 (sp. gravity)
    507 mm (20")
    0.005
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $152.38
    $152.38
    Prix promotionnel pour les nouveaux clients
    Each
       $152.38
    Supplier Number: 6605-5
  • Glass syphon, rubber tube, and clamps only
    507 mm (20")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $66.93
    $66.93
    Prix promotionnel pour les nouveaux clients
    Each
       $66.93
    Supplier Number: 6605-OSY