Synthware Round-Bottom Flasks with Four Necks
Supplier: Kemtech America
Jump to:
À propos de cet article
With identical vertical side joints.
- 3 Straight side necks
- Ground glass joints
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
13 Options Available Below
Product Details & Documents
With identical vertical side joints.
- 3 Straight side necks
- Ground glass joints
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
Product Family Options
Informations sur le produit
- CapacitéCenter Neck [ST] JointSide Neck ST JointsAvailabilityPrice
- Supplier Number: F214250
- Supplier Number: F214500
- Supplier Number: F212500
- Supplier Number: F218500
- Supplier Number: F214000
- Supplier Number: F21841L
- Supplier Number: F21842L
- Supplier Number: F21342L
- Supplier Number: F21543L
- Supplier Number: F21583L
- Supplier Number: F21845L
- Supplier Number: F21545L
- Supplier Number: F21585L