Synthware Pear-Shaped Flasks with Two Necks, Kemtech America
Supplier: Kemtech America
Jump to:
À propos de cet article
Pear-shaped with joints.
- Angled side neck
- Ground glass joint
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
- High recovery bottom
Heavy wall design.
7 Options Available Below
Product Details & Documents
Pear-shaped with joints.
- Angled side neck
- Ground glass joint
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
- High recovery bottom
Heavy wall design.
Product Family Options
Informations sur le produit
- CapacitéCenter Neck [ST] JointSide Neck ST JointsAvailabilityPrice
- Supplier Number: F341105
- Supplier Number: F341110
- Supplier Number: F341115
- Supplier Number: F341125
- Supplier Number: F341150
- Supplier Number: F341100
- Supplier Number: F341200