PYREX® Round-Bottom Boiling Flasks, Long Neck, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Round-Bottom Boiling Flasks, Long Neck, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Round-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Round-Bottom Boiling Flasks, Long Neck, Corning
5 Options Available Below
Product Details & Documents
Flasks have tooled mouths for uniform stopper fit and carefully controlled wall thicknesses to maintain proper balance between thermal expansion and mechanical strength for optimal shock resistance
They are extremely chemically durable for contaminant-free results and long life under rigid lab conditions. Working temperature: 230°C (446°F).
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89091-070
  • 29121-109
  • 89091-416
  • 89091-292
  • 29121-163
  • 89091-470
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    Rubber Stopper Size
    O.D.×H
    Availability
    Price
  • 100 ml
    2
    65×120 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-070
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 250 ml
    5
    83×149 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $45.90
    $45.90
    Prix promotionnel pour les nouveaux clients
    Case of 1
       $45.90
    Supplier Number: 4280-250
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29121-109
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 500 ml
    6
    102×203 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $675.00
    $675.00
    Prix promotionnel pour les nouveaux clients
    Case of 12
       $675.00
    Supplier Number: 4280-500
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-416
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2 L
    8
    161×308 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 18
    Case of 18 - $2,560.00
    $2,560.00
    Case of 18
       $2,560.00
    Supplier Number: 4280-2L
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-292
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 L
    8
    130×241 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $688.00
    $688.00
    Case of 12
       $688.00
    Supplier Number: 4280-1L
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29121-163
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 5 L
    11
    221×380 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $1,720.00
    $1,720.00
    Case of 6
       $1,720.00
    Supplier Number: 4280-5L
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89091-470
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®