Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
Supplier:   Kemtech America
Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
Supplier:  Kemtech America
Certificates
Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
3 Options Available Below
Product Details & Documents
Supplied with one compression cap, one Viton® O-ring, and one fluoropolymer resin-faced silicone rubber septum.
  • Threaded neck
  • Ground glass joint
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
  • Cap included
Threaded ground glass joint allows for the use of a threaded cap or a ground glass stopper.
Ordering information: For replacement caps, contact your Avantor representative.
Product Family Options
Product Information
  • Capacité
    ST Joint No.
    Availability
    Price
  • 5 ml
    14/10
    Log in for availability
    Log in for availability
    Each
    Each - $44.13
    $44.13
    New Customer Promotional Price
    Each
       $44.13
    Minimum order quantity is 1
    Supplier Number: F301005
  • 10 ml
    14/10
    Log in for availability
    Log in for availability
    Each
    Each - $50.10
    $50.10
    New Customer Promotional Price
    Each
       $50.10
    Minimum order quantity is 1
    Supplier Number: F301010
  • 25 ml
    14/10
    Log in for availability
    Log in for availability
    Each
    Each - $60.68
    $60.68
    New Customer Promotional Price
    Each
       $60.68
    Minimum order quantity is 1
    Supplier Number: F301025