Synthware Round-Bottom Flasks with Three Necks, Angled
Supplier: Kemtech America
Jump to:
À propos de cet article
Round bottom with [ST] joints.
- 3 Angled necks
- Ground glass joints
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
43 Options Available Below
Product Details & Documents
Round bottom with [ST] joints.
- 3 Angled necks
- Ground glass joints
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Product Family Options
Informations sur le produit
- CapacitéCenter Neck [ST] JointSide Neck ST JointsAvailabilityPrice
- Supplier Number: F431110
- Supplier Number: F431125
- Supplier Number: F439925
- Supplier Number: F431150
- Supplier Number: F439150
- Supplier Number: F439950
- Supplier Number: F434150
- Supplier Number: F434950
- Supplier Number: F434450
- Supplier Number: F431100
- Supplier Number: F439100
- Supplier Number: F439900
- Supplier Number: F434100
- Supplier Number: F434900
- Supplier Number: F434400
- Supplier Number: F431250
- Supplier Number: F430250
- Supplier Number: F439250
- Supplier Number: F432250
- Supplier Number: F434250
- Supplier Number: F431500
- Supplier Number: F439500
- Supplier Number: F430500
- Supplier Number: F434500
- Supplier Number: F434524
- Supplier Number: F43401L
- Supplier Number: F431045
- Supplier Number: F43242L
- Supplier Number: F43542L
- Supplier Number: F43243L
- Supplier Number: F43843L
- Supplier Number: F43293L
- Supplier Number: F43343L
- Supplier Number: F43383L
- Supplier Number: F43543L
- Supplier Number: F43583L
- Supplier Number: F43245L
- Supplier Number: F43845L
- Supplier Number: F43295L
- Supplier Number: F43345L
- Supplier Number: F43385L
- Supplier Number: F43545L
- Supplier Number: F43585L