PYREX® VISTA™ Round-Bottom Boiling Flasks, Corning | Avantor
PYREX® VISTA™ Round-Bottom Boiling Flasks, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® VISTA™ Round-Bottom Boiling Flasks, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® VISTA™ Round-Bottom Boiling Flasks, Corning
PYREX® VISTA™ Round-Bottom Boiling Flasks, Corning
4 Options Available Below
Product Details & Documents
PYREX® 7740 borosilicate glass, clear, with short neck.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These flasks have full length outer standard tapered joint. Wall thickness is controlled to provide balance between thermal expansion and mechanical strength for shock resistance.
Certifications: ASTM E-1403
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89090-308
  • 89090-328
  • 89090-352
  • 89090-368
  • 89090-390
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    ST Joint No.
    Availability
    Price
  • 100 ml
    24/40
    Discontinued
    Supplier Number: -
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-308
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 L
    24/40
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $56.40
    $56.40
    Log in pour la disponibilité
    Case of 2
    Case of 2 - $112.80
    $112.80
    Pack of 1
       $56.40
    Case of 2
       $112.80
    Supplier Number: 70320-1L
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-328
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 250 ml
    24/40
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $87.50
    $87.50
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $175.00
    $175.00
    Pack of 2
       $87.50
    Case of 4
       $175.00
    Supplier Number: 70320-250
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-352
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2 L
    24/40
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $95.00
    $95.00
    Log in pour la disponibilité
    Case of 2
    Case of 2 - $190.00
    $190.00
    Pack of 1
       $95.00
    Case of 2
       $190.00
    Supplier Number: 70320-2L
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-368
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 500 ml
    24/40
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $99.20
    $99.20
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $198.40
    $198.40
    Pack of 2
       $99.20
    Case of 4
       $198.40
    Supplier Number: 70320-500
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-390
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact