ContactUsLinkComponent
VWR® Hygrometers/Thermometers with Memory and Probe
Fournisseur:  VWR International
Certificats
undefined
VWR® Hygrometers/Thermometers with Memory and Probe
Fournisseur:  VWR International
Certificats
Spécifications
  • Catalog No:
  • 33500-098
  • 35519-050
  • Model:
  • 33500-098
  • 35519-050
  • Temperature range:
  • –18 to 93°C (0 to 199.9°F)
  • –18 to 93°C (0 to 199.9°F)
  • Temperature accuracy:
  • ±1°C (±1.8°F) from 0 to 40°C (32 to 104°F); ±2°C (±3.6°F) at ends of range
  • ±1°C (±1.8°F) from 0 to 40°C (32 to 104°F); ±2°C (±3.6°F) at ends of range
  • Temperature resolution:
  • 0.1°
  • 0.1°
  • Humidity range:
  • 10 to 95% RH
  • 10 to 95% RH
  • Humidity accuracy:
  • ±2% RH
  • ±2% RH; ±4% RH at ends of range
  • Humidity resolution:
  • 0.1%
  • 0.1%
  • Dew point range:
  • 0…50 °C (32…122 °F)
  • Dew point accuracy:
  • ±2 °C (±3.6 °F)
  • Dew point resolution:
  • 0.1°
  • Response time:
  • 30 seconds
  • 30 seconds
  • Probe:
  • 21.6L x 1.8dia. cm (81/2 x 27/32")
  • 21.6L x 1.8dia. cm (81/2 x 27/32")
  • Dimensions:
  • 18.1L x 7W x 3.2D cm (71/8 x 23/4 x 11/4")
  • 18.1L x 7W x 3.2D cm (71/8 x 23/4 x 11/4")
  • Weight:
  • 303 g (10.7 oz.)
  • 303 g (10.7 oz.)