KIMBLE® Round-Bottom Boiling Flasks, DWK Life Sciences
À propos de cet article
These threaded round bottom boiling flasks are ideal for use with [ST] 14/10 microscale apparatus.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Flasks are supplied with a blue, glass-filled nylon cap, a PTFE/silicone septum, and an ethylene propylene O-ring.
3 Options Available Below
Product Details & Documents
These threaded round bottom boiling flasks are ideal for use with [ST] 14/10 microscale apparatus.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Flasks are supplied with a blue, glass-filled nylon cap, a PTFE/silicone septum, and an ethylene propylene O-ring.
Product Family Options
Informations sur le produit
- CapacitéØ ext.AvailabilityPrice
- 5 ml25 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 1Case of 1 - $72.89$72.89Prix promotionnel pour les nouveaux clientsCase of 1$72.89Supplier Number: 746120-0005
- 10 ml31 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 1Case of 1 - $69.87$69.87Prix promotionnel pour les nouveaux clientsCase of 1$69.87Supplier Number: 746120-0010
- 25 ml42 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 1Case of 1 - $67.06$67.06Prix promotionnel pour les nouveaux clientsCase of 1$67.06Supplier Number: 746120-0025